KEY128004175 Key Organics / BIONET

5-fluoroquinazoline-2,4-diamine

  • MDL number MFCD00221785
  • Cas Number : 119584-70-2
  • Molecular Weight : 178.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 248 - 250
Iupac Name 5-fluoro-2,4-quinazolinediamine
Inchi Code 1S/C8H7FN4/c9-4-2-1-3-5-6(4)7(10)13-8(11)12-5/h1-3H,(H4,10,11,12,13)
Inchi Key ZFIDHZVBIBPRBQ-UHFFFAOYSA-N

Description

Melting Point 248 - 250
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96798

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/FA-0899_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart