KEY128004180 Key Organics / BIONET

2-(4-methylphenyl)-2-oxoethyl acetate

  • MDL number MFCD00458047
  • Cas Number : 65143-37-5
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 86 - 88
Iupac Name 2-(4-methylphenyl)-2-oxoethyl acetate
Inchi Code 1S/C11H12O3/c1-8-3-5-10(6-4-8)11(13)7-14-9(2)12/h3-6H,7H2,1-2H3
Inchi Key GMGKFZCRKFZPMC-UHFFFAOYSA-N

Description

Melting Point 86 - 88
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94456

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AB-0729_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart