KEY128004181 Key Organics / BIONET

methyl 5-fluoro-1-benzothiophene-2-carboxylate

  • MDL number MFCD07371535
  • Cas Number : 154630-32-7
  • Molecular Weight : 210.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 82
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96804

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FA-0928_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart