KEY128004186 Key Organics / BIONET

(Z)-N'-hydroxy-1H-indole-3-carboximidamide

  • MDL number MFCD00460181
  • Cas Number : 95649-37-9
  • Molecular Weight : 175.191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166 - 169
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94460

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0802_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart