KEY128004189 Key Organics / BIONET

(2E)-3-[2-(3,5-dimethylphenoxy)phenyl]prop-2-enoic acid

  • MDL number MFCD02091008
  • Cas Number : 338393-75-2
  • Molecular Weight : 268.312

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201 - 203
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65104

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1P-084_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart