KEY128004214 Key Organics / BIONET

N-methyl-5-nitropyridin-2-amine

  • MDL number MFCD00152624
  • Cas Number : 4093-89-4
  • Molecular Weight : 153.141

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178 - 180
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97200

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GA-0729_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart