KEY128004218 Key Organics / BIONET

6-(ethoxycarbonyl)pyridine-3-carboxylic acid

  • MDL number MFCD09607895
  • Cas Number : 17874-78-1
  • Molecular Weight : 195.174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >240
Purity 95
Storage temp. RT
Physical Form White to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94927

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/BB-0601_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart