KEY128004223 Key Organics / BIONET

(2E)-3-[2-(4-iodophenoxy)phenyl]prop-2-enoic acid

  • MDL number MFCD02091016
  • Cas Number : 449778-80-7
  • Molecular Weight : 366.154

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212 - 214
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68573

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-032_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart