KEY128004225 Key Organics / BIONET

1-acetyl-2,3-dihydro-1H-indole-5-sulfonamide

  • MDL number MFCD01235910
  • Cas Number : 3264-38-8
  • Molecular Weight : 240.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225 - 227
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97217

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GA-0810_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart