KEY128004235 Key Organics / BIONET

methyl (2E)-2-(4-chloro-2-methylphenoxy)-3-methoxyprop-2-enoate

  • MDL number MFCD08443943
  • Cas Number : 255908-42-0
  • Molecular Weight : 256.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72201

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3N-570S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart