KEY128004260 Key Organics / BIONET

methyl 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylate

  • MDL number MFCD05664428
  • Cas Number : 50593-91-4
  • Molecular Weight : 263.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68 - 70
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94958

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BB-0729_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart