KEY128004297 Key Organics / BIONET

1H-indole-2-carboxamide

  • MDL number MFCD00231036
  • Cas Number : 1670-84-4
  • Molecular Weight : 160.176

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232 - 243
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76167

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4P-033_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart