KEY128004299 Key Organics / BIONET

(3E)-4-[2-(4-bromo-2-fluorophenoxy)phenyl]but-3-en-2-one

  • MDL number MFCD02091019
  • Cas Number : 451485-72-6
  • Molecular Weight : 335.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83 - 85
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76182

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4P-067_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart