KEY128004309 Key Organics / BIONET

methyl 4-(3-nitropyridin-2-yl)benzoate

  • MDL number MFCD08443945
  • Cas Number : 937601-36-0
  • Molecular Weight : 258.233

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82 - 84
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77504

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Z-0708_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart