KEY128004325 Key Organics / BIONET

4-cyano-N-methoxy-N-methylbenzamide

  • MDL number MFCD07367975
  • Cas Number : 116332-64-0
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 78 - 80
Iupac Name 4-cyano-N-methoxy-N-methylbenzamide
Inchi Code 1S/C10H10N2O2/c1-12(14-2)10(13)9-5-3-8(7-11)4-6-9/h3-6H,1-2H3
Inchi Key BZGBXNBROBZKLC-UHFFFAOYSA-N

Description

Melting Point 78 - 80
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97693

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/HA-0842_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart