KEY128004339 Key Organics / BIONET

5-(prop-2-yn-1-ylsulfanyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00175753
  • Cas Number : 53918-05-1
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134 - 136
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98212

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-012C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart