KEY128004364 Key Organics / BIONET

1-(2,4-difluorophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD00033652
  • Cas Number : 6954-65-0
  • Molecular Weight : 209.152

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83 - 85
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98579

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-051C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart