KEY128004367 Key Organics / BIONET

3-hydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one

  • MDL number MFCD00017682
  • Cas Number : 6889-78-7
  • Molecular Weight : 268.268

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98640

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-057C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart