KEY128004372 Key Organics / BIONET

(2E)-1-(4-hydroxyphenyl)-3-(2,3,4-trimethoxyphenyl)prop-2-en-1-one

  • MDL number MFCD03788394
  • Cas Number : 556028-14-9
  • Molecular Weight : 314.337

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155 - 156
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98745

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-069C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart