KEY128004373 Key Organics / BIONET

(2E)-3-(4-hydroxyphenyl)-1-(naphthalen-2-yl)prop-2-en-1-one

  • MDL number MFCD03788395
  • Cas Number : 57221-63-3
  • Molecular Weight : 274.319

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98755

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-070C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart