KEY128004379 Key Organics / BIONET

(2E)-1-(4-hydroxyphenyl)-3-[4-(propan-2-yl)phenyl]prop-2-en-1-one

  • MDL number MFCD03788396
  • Cas Number : 155269-25-3
  • Molecular Weight : 266.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98841

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-079C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart