KEY128004381 Key Organics / BIONET

(2E)-3-(2-chlorophenyl)-1-(4-hydroxyphenyl)prop-2-en-1-one

  • MDL number MFCD03788397
  • Cas Number : 204974-26-5
  • Molecular Weight : 258.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98861

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-081C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart