KEY128004404 Key Organics / BIONET

1-{4-[(naphthalen-1-yl)methoxy]phenyl}ethan-1-one

  • MDL number MFCD07021293
  • Cas Number : 885267-47-0
  • Molecular Weight : 276.335

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150 - 152
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99241

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-123C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart