KEY128004405 Key Organics / BIONET

2-methylfuran-3-carbohydrazide

  • MDL number MFCD01330023
  • Cas Number : 315672-60-7
  • Molecular Weight : 140.142

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153 - 154
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99246

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-124C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart