KEY128004411 Key Organics / BIONET

1-(2-methoxy-5-nitrophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD00523329
  • Cas Number : 17392-67-5
  • Molecular Weight : 248.194

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158 - 159
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99306

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-131C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart