KEY128004418 Key Organics / BIONET

5-(3-chlorophenyl)-4-(2-methylpropyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD05150471
  • Cas Number : 26028-64-8
  • Molecular Weight : 267.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99427

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-145C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart