KEY128004434 Key Organics / BIONET

4-(cyclohexylsulfanyl)-3-nitrobenzaldehyde

  • MDL number MFCD00098772
  • Cas Number : 270262-95-8
  • Molecular Weight : 265.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 99
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99633

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-168C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart