KEY128004441 Key Organics / BIONET

2-[(4-aminophenyl)sulfanyl]-N-[4-(trifluoromethoxy)phenyl]acetamide

  • MDL number MFCD05155029
  • Cas Number : 763126-91-6
  • Molecular Weight : 342.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99728

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-178C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart