KEY128004447 Key Organics / BIONET

N-[(2H-1,3-benzodioxol-5-yl)methyl]-3-nitroaniline

  • MDL number MFCD00422251
  • Cas Number : 191595-08-1
  • Molecular Weight : 272.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99 - 100
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99848

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-190C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart