KEY128004451 Key Organics / BIONET

(2,4-dichlorophenyl)thiourea

  • MDL number MFCD00022161
  • Cas Number : 6326-14-3
  • Molecular Weight : 221.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 163 - 165
Iupac Name N-(2,4-dichlorophenyl)thiourea
Inchi Code 1S/C7H6Cl2N2S/c8-4-1-2-6(5(9)3-4)11-7(10)12/h1-3H,(H3,10,11,12)
Inchi Key HSWCDFOASWNGOM-UHFFFAOYSA-N

Description

Melting Point 163 - 165
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99890

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-194C_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart