KEY128004459 Key Organics / BIONET

1-(4-bromophenyl)-3-[4-(3-chlorophenyl)piperazin-1-yl]propan-1-one

  • MDL number MFCD05261306
  • Cas Number : 952182-60-4
  • Molecular Weight : 407.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73 - 74
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100003

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-205C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart