KEY128004469 Key Organics / BIONET

2-(4-fluorophenyl)-4H-3,1-benzoxazin-4-one

  • MDL number MFCD00087177
  • Cas Number : 18600-51-6
  • Molecular Weight : 241.221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180 - 181
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100145

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-219C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart