KEY128004503 Key Organics / BIONET

3-bromo-1-methyl-5-nitro-1H-indazole

  • MDL number MFCD09607930
  • Cas Number : 74209-25-9
  • Molecular Weight : 256.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 220 - 222
Iupac Name 3-bromo-1-methyl-5-nitro-1H-indazole
Inchi Code 1S/C8H6BrN3O2/c1-11-7-3-2-5(12(13)14)4-6(7)8(9)10-11/h2-4H,1H3
Inchi Key QWSNHRDLAUTWJP-UHFFFAOYSA-N

Description

Melting Point 220 - 222
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95603

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CB-0820_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart