KEY128004507 Key Organics / BIONET

2-(4-methoxyphenyl)-1,3-thiazolidine

  • MDL number MFCD00222821
  • Cas Number : 31404-08-7
  • Molecular Weight : 195.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 100 - 102
Iupac Name 2-(4-methoxyphenyl)-1,3-thiazolidine
Inchi Code 1S/C10H13NOS/c1-12-9-4-2-8(3-5-9)10-11-6-7-13-10/h2-5,10-11H,6-7H2,1H3
Inchi Key OQFUUZFCTONCOU-UHFFFAOYSA-N

Description

Melting Point 100 - 102
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100828

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/KA-0742_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart