KEY128004511 Key Organics / BIONET

tert-butyl 4-[4-(methoxycarbonyl)-2-oxopyrrolidin-1-yl]piperidine-1-carboxylate

  • MDL number MFCD08689666
  • Cas Number : 937601-48-4
  • Molecular Weight : 326.393

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 100
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87969

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7W-0612_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart