KEY128004534 Key Organics / BIONET

(E)-N-[(1H-pyrrol-2-yl)methylidene]hydroxylamine

  • MDL number MFCD00965095
  • Cas Number : 32597-34-5
  • Molecular Weight : 110.116

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169 - 171
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90206

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8P-018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart