KEY128004564 Key Organics / BIONET

1-cyclopropylpiperidin-4-one

  • MDL number MFCD07374375
  • Cas Number : 62813-01-8
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 55 @ 2 mbar
Iupac Name 1-cyclopropyl-4-piperidinone
Inchi Code 1S/C8H13NO/c10-8-3-5-9(6-4-8)7-1-2-7/h7H,1-6H2
Inchi Key DTUJRJIWGWTNFQ-UHFFFAOYSA-N

Description

Melting Point 55 @ 2 mbar
Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101702

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/LA-0813_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart