KEY128004566 Key Organics / BIONET

2-(benzyloxy)ethan-1-amine

  • MDL number MFCD00235180
  • Cas Number : 38336-04-8
  • Molecular Weight : 151.209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 97
Storage temp. Store under N2
Physical Form Colourless to Off-white Liquid
Shipping Temperature Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101707

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P410,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/LA-0829_SDS.pdf
pictograms GHS05,GHS07
hstatements H302,H312,H314,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart