KEY128004595 Key Organics / BIONET

6-methoxy-1H-indole-3-carbaldehyde

  • MDL number MFCD03848058
  • Cas Number : 70555-46-3
  • Molecular Weight : 175.187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186 - 188
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102078

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MA-0823_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart