KEY128004599 Key Organics / BIONET

2-hydroxy-1,3-benzothiazole-6-carboxylic acid

  • MDL number MFCD08444018
  • Cas Number : 99615-68-6
  • Molecular Weight : 195.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96030

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/DA-0926_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart