KEY128004630 Key Organics / BIONET

3-methyl-2-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)pentanoic acid

  • MDL number MFCD04367477
  • Cas Number : 180923-81-3
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 135 - 138
Iupac Name 3-methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid
Inchi Code 1S/C14H17NO3/c1-3-9(2)12(14(17)18)15-8-10-6-4-5-7-11(10)13(15)16/h4-7,9,12H,3,8H2,1-2H3,(H,17,18)
Inchi Key SDNDNHOVHJTJPB-UHFFFAOYSA-N

Description

Melting Point 135 - 138
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106653

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/NA-0808_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart