KEY151861539 Key Organics / BIONET

ethyl 3-(trifluoromethyl)pyridine-2-carboxylate

  • MDL number MFCD09864691
  • Cas Number : 952182-76-2
  • Molecular Weight : 219.163

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7397

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11Z-0838_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart