KEY151861540 Key Organics / BIONET

({6-bromoimidazo[1,2-a]pyridin-3-yl}methyl)dimethylamine hydrochloride

  • MDL number MFCD09817424
  • Cas Number : 1134570-17-4
  • Molecular Weight : 290.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223 - 225DEC
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88298

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7Z-0808_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart