KEY151861542 Key Organics / BIONET

6-chloro-1,3-dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD09817432
  • Cas Number : 952183-47-0
  • Molecular Weight : 249.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49 - 51
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95619

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/CB-0857_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart