KEY151861550 Key Organics / BIONET

2-[4-(trifluoromethyl)piperidin-1-yl]-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD09864683
  • Cas Number : 1000339-88-7
  • Molecular Weight : 264.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 42 - 45
Iupac Name 2-[4-(trifluoromethyl)-1-piperidinyl]-1,3-thiazole-5-carbaldehyde
Inchi Code 1S/C10H11F3N2OS/c11-10(12,13)7-1-3-15(4-2-7)9-14-5-8(6-16)17-9/h5-7H,1-4H2
Inchi Key DGADTZIGHLQBTO-UHFFFAOYSA-N

Description

Melting Point 42 - 45
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100876

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/KB-0721_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart