KEY151861559 Key Organics / BIONET

ethyl 1-({[(tert-butoxy)carbonyl]amino}sulfonyl)piperidine-4-carboxylate

  • MDL number MFCD09864838
  • Cas Number : 1000018-25-6
  • Molecular Weight : 336.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79 - 81
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100897

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KB-0815_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart