KEY151861561 Key Organics / BIONET

tert-butyl 4-[5-bromo-3-(methoxycarbonyl)pyridin-2-yl]piperazine-1-carboxylate

  • MDL number MFCD09864832
  • Cas Number : 1000018-22-3
  • Molecular Weight : 400.273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122 - 124
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101717

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LB-0703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart