KEY151861571 Key Organics / BIONET

tert-butyl 4-(3-ethoxy-3-oxopropanoyl)piperidine-1-carboxylate

  • MDL number MFCD04115569
  • Cas Number : 479630-08-5
  • Molecular Weight : 299.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-(3-ethoxy-3-oxopropanoyl)-1-piperidinecarboxylate
Inchi Code 1S/C15H25NO5/c1-5-20-13(18)10-12(17)11-6-8-16(9-7-11)14(19)21-15(2,3)4/h11H,5-10H2,1-4H3
Inchi Key CWTULLANCDLMGK-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Low Melting Solid,Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96822

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/FB-0733_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart