KEY151861574 Key Organics / BIONET

2-hydroxy-2-(4-propoxyphenyl)acetic acid

  • MDL number MFCD07368282
  • Cas Number : 79694-16-9
  • Molecular Weight : 210.229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96825

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FB-0736_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart