KEY151861586 Key Organics / BIONET

methyl 2-amino-2-(3,4-dichlorophenyl)acetate hydrochloride

  • MDL number MFCD04115512
  • Cas Number : 1078611-21-8
  • Molecular Weight : 270.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97295

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GB-0722_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart